missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VETRANAL™ Nafcillin sodium salt, MilliporeSigma™ Supelco™
Description
Pharmaceutical (small molecule)
Specifications
Specifications
| Product Line | VETRANAL |
| CAS | 985-16-0 |
| Physical Form | Neat |
| Linear Formula | C21H21N2 NaO5 S |
| Synonym | (2 S,5 R,6 R)-6-{[(2-Ethoxy-1-naphthalenyl)carbonyl]amino}-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0 ]heptane-2-carboxylic acid sodium salt; 6-(2-Ethoxy-1-naphthamido)penicillin sodium salt |
| Type | Analytical Standard |
| Quantity | 100 mg |
| Formula Weight | 436.46 |
| Grade | Analytical Standard |
VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH