missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VETRANAL™ Monensin sodium salt hydrate, MilliporeSigma™ Supelco™
Description
Cleaning Products, Cosmetics, Food and Beverages, Personal Care and Pharmaceutical (small molecule)
Specifications
Specifications
| Product Line | VETRANAL |
| CAS | 22373-78-0 |
| Physical Form | Neat |
| Linear Formula | C36H61 NaO11 · xH2 O |
| Type | Analytical Standard |
| Quantity | 100 mg |
| Formula Weight | 692.85 (anhydrous basis) |
| Grade | Analytical Standard |
VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH