missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VETRANAL™ Metronidazole-13C2,15N2, MilliporeSigma™ Supelco™
Description
Pharmaceutical (small molecule)
Specifications
Specifications
| Product Line | VETRANAL |
| CAS | 1173020-03-5 |
| Physical Form | Neat |
| Linear Formula | C413C2H9N15N2O3 |
| MDL Number | MFCD16652531 |
| Synonym | 2-(Methyl-13 C)-5-nitro(imidazole-2-13 C-1,3-15N2)-1-ethanol |
| Type | Analytical Standard |
| Quantity | 10 mg |
| Formula Weight | 175.13 |
| Grade | Analytical Standard |
VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH