missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VETRANAL™ Etoricoxib, MilliporeSigma™ Supelco™
Description
Pharmaceutical (small molecule)
Specifications
Specifications
| Product Line | VETRANAL |
| CAS | 202409-33-4 |
| Physical Form | Neat |
| Linear Formula | C18H15 ClN2O2 S |
| MDL Number | MFCD06797512 |
| Synonym | 5-Chloro-3-[4-(methylsulfonyl)phenyl]-2-(2-methyl-5-pyridinyl)pyridine; 5-Chloro-6 ′-methyl-3-[4-(methylsulfonyl)phenyl]-2,3 ′-bipyridine |
| Type | Analytical Standard |
| Quantity | 25 mg |
| Formula Weight | 358.84 |
| Grade | Analytical Standard |
VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH