missing translation for 'onlineSavingsMsg'
Learn More

Kent Scientific VetFlo™ Traditional Anesthesia System with Single Channel Stand

Catalog No. p-7077923
Encompass
Change view
Click to view available options
Flow Rate:
0 to 1 L/min.
0.2 to 4 L/min.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Flow Rate
13005201 0 to 1 L/min.
13005202 0.2 to 4 L/min.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 13005201 Supplier Kent Scientific Supplier No. VETFLO1205S

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Stand, inlet/outlet caps, filter canister, O2 tubing and adapter

For laboratory, veterinary anesthesia delivery. Kent Scientific VetFlo™ Traditional Anesthesia System with Single Channel Stand is self contained and portable.

  • Convenient tabletop design, self-contained for efficiency
  • Station specific delivery for precise control
  • Y-adapter tubing for easy transfer from induction chamber to anesthesia mask
  • Individually controlled multiple stations
  • Easy follow chart
  • Different flow rate of anesthesia for each station

Warranty

One year (parts, labor charges), Three year (vaporizer)

Specifications

For Use With (Application) Anesthesia, Animal Surgery
Type High Flow Vaporizer
Flow Rate 0 to 1 L/min.
Dimensions (D x W x H) Stand: 25.4 x 30.48 x 33.02 cm (10 x 12 x 13 in.)