Learn More
STERIS Vesta-Syde™ SQ st Quaternary Ammonium Disinfectant One Solution WFI
This SQ st Quaternary Ammonium Disinfectant One Solution WFI is an excellent fungicidal sterile concentrate in a 1 oz. pouch packaged with 1 gal. WFI.
$1,059.02
Description
STERIS™ Vesta-Syde™ SQ st Quaternary Ammonium Disinfectant is the sterile version of the Vesta-Syde™ SQ family of disinfectants made specifically for use on hard non-porous surfaces. The Vesta-Syde™ SQ products are all made with the same quaternary ammonium formulation, but are processed and packaged differently for specific applications. At a use-dilution rate of 0.8% v/v (1:128), the Vesta-Syde™ SQ products kill a broad spectrum of common bacteria and fungi without harming the surface.
How Vesta-Syde SQ st Disinfectant Works
STERIS™ Vesta-Syde™ st SQ Quaternary Ammonium Disinfectant is a liquid concentrate specifically formulated for disinfection of hard surfaces. The use-dilution can be applied manually or sprayed.
Why Vesta-Syde SQ st Disinfectant?
- Flexibility
- Excellent fungicidal efficacy can support reduced sporicide use to control mold contamination
- Convenience
- Gamma irradiated and double bagged for easy introduction into your aseptic areas
- Productivity
- Formulated with biodegradable surfactants that provide excellent cleaning ability
- Availability
- Active Ingredients notified according to the Biocide Product Directive and registered with the US EPA
- Assurance
- Each lot documented by a Certificate of Analysis including irradiation and sterility results