Learn More
Texwipe™ Vertex™ Sterile Dry Cleanroom Wipers
These dry wipers are designed for high sorption capacity, thereby reducing the number of wipes needed for spill control. The sealed edge minimizes particle release from the wiper edges.
Supplier: Texwipe™ TX3221
Description
Vertex™ Sterile Dry Cleanroom Wipers are designed for high sorption capacity to reduce the number of wipes needed for spill control. The sealed edge minimizes particle release from the wiper edges.
- Low levels of ions, NVRs (nonvolatile residues), particles and fibers (low linting wipes)
- Designed for high sorption capacity- ideal for spill control, cleaning and solution applications
- Tested for cytotoxicity and bacterial endotoxins for extra level of contamination control assurance
- Meets USP 797 and USP 800 requirements
- 100% polyester knit
Specifications
| Polyester | |
| ISO Class 3 to 7, Class 1 to 10,000 | |
| Sealed | |
| Sterile |
| Autoclavable | |
| White | |
| Critical Cleaning | |
| Grade A to D |