missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ VersiCap Mat, 96-well, ultra clear cap strips

Flexible mats of flat, domed cap strips for PCR or ultra clear cap strips for qPCR applications for use with Thermo Scientific 96-well PCR plates and strip tubes.

$221.04 - $987.10

Specifications

Color Clear
Material Polypropylene
No. of Wells 96
Format Strips of 8 Optical Caps
Product Type PCR Plate Cap Strip
View More Specs

Products 2
Catalog Number Mfr. No. No. per Case Price Quantity & Availability  
Catalog Number Mfr. No. No. per Case Price Quantity & Availability  
AB1820
View Documents Promotion Details
Thermo Scientific™
AB1820
25
Case for $221.04
Only null left
 
AB1820100
View Documents Promotion Details
Thermo Scientific™
AB1820100
100
Case for $987.10
Only null left
 
Description

Description

Thermo Scientific VersiCap Mats are unique, with each strip of eight caps being linked to each other forming the familiar 12 x 8 or 96-well ANSI format. The small tear points between strips enable a single strip or as many as required for the individual experiment to be separated.

The Thermo Scientific VersiCap Mats are unique, with each strip of eight caps being linked to each other forming the familiar 12 x 8 or 96-well ANSI format. The small tear points between strips enable a single strip or as many as required for the individual experiment to be separated.

Highlights

  • Multiple cap strips can be applied at the same time when sealing qPCR plates, thereby reducing setup time
  • Remove the need to cut up adhesive seals
  • Suitable for use with all Thermo Scientific 96-well PCR plates as well as standard and low profile strip tubes
Applications
  • PCR
  • qPCR/Real-Time PCR

Autoclavable Resistance: Successfully Tested
DMSO (100%) Resistance: Successfully Tested
Ethanol Resistance: Successfully Tested
Gamma Irradiation Resistance: Successfully Tested
Hazardous: No
Isopropanol Resistance: Successfully Tested
Long Term Storage: Successfully Tested
PCR (including waterbath): Successfully Tested
Peelable: Successfully Tested
Pierceable: Not Recommended
qPCR:Ultra Clear caps: Successfully Tested
Domed or Flat caps: Not Recommended
Resealable: Successfully Tested
Sealing Temperature Range: −20°C to +120°C
Shelf Life: 60 Months
Shipping Condition: Ambient
Storage Condition: Ambient
UV Irradiation Resistance: Successfully Tested

  • PCR
  • qPCR/Real-Time PCR

Certifications

Certified free from human DNA, RNases, and DNases

Specifications

Specifications

Clear
96
PCR Plate Cap Strip
Yes
96-well Plate
Polypropylene
Strips of 8 Optical Caps
VersiPlate
-20°C to +120°C
Videos
SDS
Documents

Documents