missing translation for 'onlineSavingsMsg'
Learn More

Honeywell™ VeriShield™ 100 Series Over-the-Head Dielectric Ear Muffs

Catalog No. 19085530
Change view
Click to view available options
CSA Z94.2:
Class A
Class AL
Foldable:
No
Noise Reduction Rating (NRR):
23 dB
26 dB
29 dB
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. CSA Z94.2 Foldable Noise Reduction Rating (NRR)
19085530 Class AL No 29 dB
19085525 Class A No 23 dB
19085528 Class A No 26 dB
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19085530 Supplier Honeywell™ Supplier No. 1035195VS

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

VeriShield™ Over-the-Head Dielectric Ear Muffs feature a durable, non-deforming plastic headband and are ideal for electrical environments.

  • Dielectric suitable for electrical environments
  • Durable plastic headband
  • Steel-wire construction with lightweight cushioned headband and precision micro adjustments are designed for excellent fit around the ear
  • Memory foam ear cushions combine dense and soft foam
  • Reduces squeezing pressure on the head and enhances comfort
  • Offered in low level, mid level or high level models

Specifications

ANSI S3.1974 Yes
CSA Z94.2 Class AL
Cushion Color Black
For Use With (Application) Construction, Electrical, Manufacturing, Oil and Gas, Mining, Steel and Metals, Agriculture
High Visibility No
Model VS130D
Pairs per Package 1 Ea.
Noise Reduction Rating (NRR) 29 dB
Product Type VeriShield
Headband Color Black
Dielectric Yes
Style Over-the-head
Foldable No
Show More Show Less