missing translation for 'onlineSavingsMsg'
Learn More

Honeywell™ VeriShield™ 100 Series Over-the-Head Dielectric Ear Muffs

Catalog No. 19085525
Change view
Click to view available options
CSA Z94.2:
Class A
Class AL
Foldable:
No
Noise Reduction Rating (NRR):
23 dB
26 dB
29 dB
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. CSA Z94.2 Foldable Noise Reduction Rating (NRR)
19085525 Class A No 23 dB
19085528 Class A No 26 dB
19085530 Class AL No 29 dB
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19085525 Supplier Honeywell™ Supplier No. 1035190VS

VeriShield™ Over-the-Head Dielectric Ear Muffs feature a durable, non-deforming plastic headband and are ideal for electrical environments.

  • Dielectric suitable for electrical environments
  • Durable plastic headband
  • Steel-wire construction with lightweight cushioned headband and precision micro adjustments are designed for excellent fit around the ear
  • Memory foam ear cushions combine dense and soft foam
  • Reduces squeezing pressure on the head and enhances comfort
  • Offered in low level, mid level or high level models

Specifications

ANSI S3.1974 Yes
CSA Z94.2 Class A
Cushion Color Black
For Use With (Application) Construction, Electrical, Manufacturing, Oil and Gas, Mining, Steel and Metals, Agriculture
High Visibility No
Model VS110D
Pairs per Package 1 Ea.
Noise Reduction Rating (NRR) 23 dB
Product Type VeriShield
Headband Color Black
Dielectric Yes
Style Over-the-head
Foldable No
Show More Show Less