missing translation for 'onlineSavingsMsg'
Learn More

Versenate Solution, Reagents

Catalog No. 01335344
Change view
Click to view available options
Quantity:
20 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01335344 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01335344 Supplier Labchem Inc Supplier No. CE25120020F

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Versenate Solution, 1 mL = 1 mg CaCO3, Reagent, use for Metals analysis (EDTA method). Manufactured in ISO 9001 facility. NIST Traceable.

Specifications

Color Colorless
Packaging Type Cubitainer
Quantity 20 L
For Use With (Application) Metals Analysis (EDTA method)
Form Liquid