missing translation for 'onlineSavingsMsg'
Learn More

3M™ Versaflo™ S-Series High Durability Hood

Catalog No. p-200087363
Change view
Click to view available options
Model:
S-533 L
S-533 S
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model
19805600 S-533 S
19805599 S-533 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19805600 Supplier 3M™ Supplier No. 7000127432
Only null left

The 3M™ S-Series High Durability Hood S-500 Series with integrated head suspension and face seal is similar to the S-400 Series but with a more durable, soft, quiet, low-linting fabric that more readily drapes comfortably over the wearer.

  • Integrated head suspension provides head, face, neck and shoulder coverage
  • Protects face and eyes from liquid splash and low energy flying particles and dust (Grade F)
  • Manufactured using durable, soft, low-linting fabric for user comfort
  • Suitable for applications which require frequent replacement of the entire headtop assembly or those where there is less soiling of the outer fabric
  • No assembly required; ready to use straight out of the box
  • Weighs 170 g
  • CE approved and rated to EN 12941 TH3, EN 166:2:F:3 and EN14594 3A
  • Head, Face, Neck and Shoulder coverage
  • White

Specifications

Assigned Protection Factor (APF) 1000
Certifications/Compliance NIOSH
Color White
Material Polyurethane-coated Knitted Polyamide
Product Type Hood with Faceseal
Size Small/Medium
For Use With (Application) Assembly, facility maintenance, machine operation, painting, sanding
For Use With (Equipment) GVP, Breathe-Easy™, V Series, T-300, T-600, T-800
Material (Lens) Coated Polycarbonate
Model S-533 S
Quantity 1 Ea.
Dimensions (L x W x H) 10 x 9.63 x 8.19 in. (25.4 x 24.46 x 20.8 cm)
Show More Show Less