missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ Versaflo™ S-Series Headcovers and Hoods
Description
- S-Series respiratory headcovers and hoods are specifically designed to work together with your Versaflo™ Powered Air Respirator and 3M™ Supplied Air Respirator Systems.
- Provides head, face, neck and shoulder coverage from dust and paint spray.
- Provides a good field of view.
- Helps protect eyes and face from liquid splash and low energy flying particles and dust.
- Versatile accessories available that can help you adapt the hood to your particular environment.
Specifications
Specifications
| Assigned Protection Factor (APF) | 1000 |
| Certifications/Compliance | NIOSH |
| Description | Premium Replacement Hood with Double Shroud, S-657 |
| Color | White |
| Material | Polypropylene |
| Product Type | Hood with Inner Shroud |
| For Use With (Application) | Assembly, facility maintenance, machine operation, painting, sanding |
| For Use With (Equipment) | 3M™ powered air purifying respirator and supplied air respirator valves |
| Includes | S-607 Hood with Inner Shroud, S-950 Premium Head Suspension, S-955 Airflow Adjustment Cover, S-954 Size Reducing Pad, S-922 Peel-Off |
| Material (Lens) | PETG |
| Show More |