Learn More
DuPont™ Tychem™ 10000 Encapsulated Level B
10000 Encapsulated Level B offers a high level of protection against toxic, corrosive gases and liquid chemicals
Supplier: DuPont™ TK527TLYLG000100
Description
- 10000 Encapsulated Level B design provides protection from liquid splash exposure for both the wearer and respiratory equipment
- Integrated socks composed of garment material
- Attached flaps are designed to cover boot tops to help reduce potential for liquid intrusion
- Manufactured under specifications that do not contain natural rubber latex
- Made in the USA, North American Free Trade Agreement (NAFTA) compliant and Trade Agreement Act (TAA) compliant
- Also provides coverage from overhead liquid splash (does not provide vapor protection)
- Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing.
- Standard 40 mil PVC faceshield
- Front entry design allows wearer to partially don garment (maintain standby position) without assistance
- Expanded back to accommodate self-contained air breathing apparatus (SCBA)
- Liquid resistant, extra long zipper to increase garment opening and aid donning and doffing
- Storm flap covers zipper which can be secured by the wearer with rugged hook and loop material to prevent intrusion at zipper
- Elastic opening for tighter fit at wrist
Specifications
| Lime Yellow | |
| Large | |
| Zipper |
| Tychem | |
| Boot Overflaps | |
| Taped |