missing translation for 'onlineSavingsMsg'
Learn More

3M™ Versaflo™ Peel-Off Visor Cover

Catalog No. 19805623
Change view
Click to view available options
Size:
Small
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19805623 Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19805623 Supplier 3M™ Supplier No. 7000127435

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

3M™ Versaflo™ Peel-Off Visor Cover is a self-adhesive cover that protects the visor from contaminants and scratches.

  • For use with 3M™ Versaflo™ S-600, S-700 and S-800 head covers and hoods
  • Self-adhesive visor covers can be easy applied and torn off
  • Protects the visor from contaminants and scratches

Specifications

Color Transparent
Product Type Visor Cover
Size Small
For Use With (Equipment) 3M™ Versaflo™ S-600, S-700, and S-800 head covers and hoods