missing translation for 'onlineSavingsMsg'
Learn More

3M™ Versaflo™ PAPR System Filters and Cartridges

Catalog No. p-4186951
Change view
Click to view available options
Product Type:
Cartridge
HE Filter
Filter Type:
Combination HE Particulate/Acid Gas/HEPA
Combination HE Particulate/Ammonia/Methylamine
Combination HE Particulate/Formaldehyde
Combination HE Particulate/Organic Vapor/Acid Gas/HEPA
Combination HE Particulate/Organic Vapor/HEPA
Particulate
Color:
Black/Magenta
Magenta
Magenta/Green
Magenta/Olive
Magenta/White
Magenta/Yellow
For Use With (Application):
Healthcare, Pharmaceutical, Heavy Industrial, Manufacturing, General Manufacturing
Pharmaceutical
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Filter Type Color For Use With (Application)
19268007 HE Filter Particulate Magenta Healthcare, Pharmaceutical, Heavy Industrial, Manufacturing, General Manufacturing
19268012 Cartridge Combination HE Particulate/Acid Gas/HEPA Magenta/White Pharmaceutical
19268010 Cartridge Combination HE Particulate/Organic Vapor/HEPA Black/Magenta Pharmaceutical
19268011 Cartridge Combination HE Particulate/Organic Vapor/Acid Gas/HEPA Magenta/Yellow Pharmaceutical
19268013 Cartridge Combination HE Particulate/Ammonia/Methylamine Magenta/Green Pharmaceutical
19268014 Cartridge Combination HE Particulate/Formaldehyde Magenta/Olive Pharmaceutical
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 19268007 Supplier 3M™ Supplier No. 7100028247
Only null left

Compatible with powered air purifying respirators in the TR-600 Series

  • Filters and cartridges work with belt-mounted Versaflo PAPR units
  • Recommended for use in healthcare, pharmaceutical, heavy industrial, manufacturing and/or general manufacturing environments
  • NIOSH approved
  • 3M Catalog# TR-6320N, TR-6350N, TR-6360N, TR-6510N, TR-6530N, TR-6710N-5, TR-6820N

Compliance

NIOSH approved.

Specifications

Certifications/Compliance NIOSH
Color Magenta
Filter Type Particulate
Product Type HE Filter
For Use With (Application) Healthcare, Pharmaceutical, Heavy Industrial, Manufacturing, General Manufacturing
For Use With (Equipment) TR-600 PAPR
Model TR-6710N-5
Quantity 10/Pk.