missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ Versaflo™ PAPR System Filters and Cartridges
Description
- Filters and cartridges work with belt-mounted Versaflo PAPR units
- Recommended for use in healthcare, pharmaceutical, heavy industrial, manufacturing and/or general manufacturing environments
- NIOSH approved
- 3M Catalog# TR-6320N, TR-6350N, TR-6360N, TR-6510N, TR-6530N, TR-6710N-5, TR-6820N
Compliance
NIOSH approved.
Specifications
Specifications
| Certifications/Compliance | NIOSH |
| Color | Magenta/Yellow |
| Filter Type | Combination HE Particulate/Organic Vapor/Acid Gas/HEPA |
| Product Type | Cartridge |
| For Use With (Application) | Pharmaceutical |
| For Use With (Equipment) | TR-600 PAPR |
| Model | TR-6530N |
| Quantity | 10/Pk. |