Learn More
3M™ Versaflo™ PAPR Rechargeable Battery Power Supply Devices
Catalog No. 18783b
Change view
Click to view available options
For Use With (Equipment):
Breathe Easy PAPR Systems
Versaflo TR-300 or Speedglas TR-300-SG PAPRs
Product Type:
Battery Port Plug
Multi-Unit Charging Station
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Product Type
18783B Breathe Easy PAPR Systems Multi-Unit Charging Station
19039568 Versaflo TR-300 or Speedglas TR-300-SG PAPRs Multi-Unit Charging Station
29663289 - Battery Port Plug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18783B Supplier 3M™ Supplier No. 520 03 72
Only null left
Choice of single or multiple unit power supply sources
  • Provide extended use of powered air purifying respirator systems
  • 3M Catalog# GVP-110, GVP-210, TR-341N, TR-344N, TR-641N, TR-644N, TR-941N, TR-944N, 520-04-24
Compliance
NIOSH approved.
Specifications
Product Type Multi-Unit Charging Station
For Use With (Equipment) Breathe Easy PAPR Systems
No. of Ports Five