missing translation for 'onlineSavingsMsg'
Learn More

3M™ Versaflo™ M-Series Helmet Assembly with Speedglas™ Welding Shield

Catalog No. 19070935
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19070935 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19070935 Supplier 3M™ Supplier No. 7100168249

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

3M™ Versaflo™ M-Series Helmet Assembly with Speedglas™ Welding Shield for use with Speedglas™ 9100 Series Auto Darkening Filter.

  • Welding shield kit
  • Available as replacement parts
  • Weld shield can be installed with common hand held tools
  • Weld shield kit compatible with only existing M-Series headtops
  • User is able to lift weld shield independently from respiratory visor
  • Able to lift visors in non-hazardous areas to help facilitate communications
  • Compatible with 9100XXi ADF with Natural Color Technology
  • 3M Catalog# M-153SG

Specifications

Certifications/Compliance ANSI Z89.1 Type I, Class G
Color Black
For Use With (Equipment) 3M M-407 Helmet Configurations, Speedglas 9100 Series Auto-darkening Filters
For Use With (Application) Welding
Model M-153SG
Product Type Headgear
Quantity 1 Ea.
Size Standard
Visor Type Clear