Learn More
3M™ Versaflo™ High Temperature Breathing Tube Cover, BT-926
Catalog No. 296366704
Change view
Click to view available options
Material:
Nomex
Catalog No. Material
296366704 Nomex
1 options
Catalog No. 296366704 Supplier 3M™ Supplier No. 7000052729
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Used to help protect the breathing tube when used in high temperature environments.
  • Help prolong the life of your breathing tube by fitting it with high temperature protection
  • 3M produces a wide variety of breathing tubes and powered air purifying respirators (PAPR)
  • In addition to powered systems, 3M also offers supplied air respirators (SAR) using air from an air tank or a different external source
Specifications
Material Nomex
Product Type Breathing Tube
For Use With (Application) Abrasive blasting
Model BT-926
Quantity 1/Cs.