missing translation for 'onlineSavingsMsg'
Learn More

3M™ Versaflo™ Headcover PAPR Kit

Catalog No. p-7155906
Change view
Click to view available options
Size:
Large/X-Large
Small/Medium
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19121501 Small/Medium
19121502 Large/X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19121501 Supplier 3M™ Supplier No. 7100100424
Only null left

Headcover PAPR kit offers a complete powered air respirator system that includes all necessary components to get started.

  • Cover your bases - Utilizes cartridges to protect against gas and organic vapors, combined with filters for protection against particulates.
  • Get more out of your workday - Longer run time, reduced downtime with a high capacity battery and charge-level indicators to keep you prepared and informed.
  • A broader view - See more of your work world with this convenient, maintenance-free disposable headcover.
  • Keeping it clean - Fully submersible product features a smooth surface design that’s quick and easy to clean and decontaminate.
  • Gas and Vapor protection can be provided with cartridges not part of this kit
  • System provides integrated solutions for respiratory protection, with limited eye, head, face and skin protection as well
  • Includes: Headcover, TR-602N PAPR Unit, TR-630 Standard Battery, BT-30 Length Adjusting Breathing Tube, TR-627 Easy Clean Belt, TR-971 Airflow Indicator, TR-6712N (HE) Particulate Filter, TR-6700FC Filter Cover, TR-641N Single Station Battery Charger, TR-653 Cleaning and Storage Kit, TR-991 Storage Bag
  • 3M Catalog# TR-600-HKL , 3M Catalog# TR-600-HKS

Specifications

Battery Life 4 to 12 hours
Battery Type Lithium-ion
Cartridge Type Gas and Organic Vapors (sold separately)
Certifications/Compliance NIOSH Approved
Color Gray
Filter Type HEPA
Product Type PAPR Healthcare Systems and Kits
Size Small/Medium
For Use With (Application) Pharmaceutical and Healthcare
Model TR-600-HKS