missing translation for 'onlineSavingsMsg'
Learn More

3M™ Versaflo™ Backpack Adaptor

Catalog No. 296366456
Change view
Click to view available options
Color:
Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
296366456 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 296366456 Supplier 3M™ Supplier No. 7100035843

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

3M™ Versaflo™ Backpack Adaptor helps fit the turbo unit to the backpack harness, which allows it to be worn over the shoulders.

  • Adaptor helps fit the turbo unit to the backpack harness, so it can be worn over the shoulders
  • Easy to connect to turbo unit
  • For use with the 3M™ Versaflo™ BPK-01 backpack

Specifications

Color Black
Product Type Backpack Adapter
For Use With (Equipment) 3M™ Versaflo™ Powered Air Turbo TR-600 and BPK-01 Backpack