Learn More
3M™ Versaflo™ Airflow Indicator, TR-971
Catalog No. 296366451
Change view
Click to view available options
1/Pk.
Catalog No.
296366451 1/Pk.
1 options
Catalog No. 296366451 Supplier 3M™ Supplier No. 70071668076
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
May be used to confirm air flow levels of the 3M™ Versaflo™ Powered Air Purifying Respirator TR-600 Series. It is designed to enable testing at various elevations and temperatures within the operational range of the PAPR system.
  • Delve into 3M™ Products and experience the wide variety of products and replacement parts offered with powered air purifying respirators (PAPR)
  • In addition to powered systems, 3M also offers supplied air respirators (SAR) using air from an air tank or a different external source
Specifications
Product Type Air Flow Indicator
For Use With (Application) Used in applications such as assembly, blasting, cleaning, metal repair, painting
Quantity 1/Pk.