Learn More
3M™ Versaflo™ Air Flow Indicator TR-973
Catalog No. 19070958
Change view
Click to view available options
Quantity: 1
Catalog No.
19070958 Quantity: 1
1 options
Catalog No. 19070958 Supplier 3M™ Supplier No. 7100167216
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Designed to enable testing at various elevations and temperatures within the operational range of the PAPR system. May be used to indicate air flow levels of the TR-600 and TR-800 series PAPRs when used with tight fitting face pieces.
  • Enables confirmation of airflow
  • Confirms factory calibration of airflow
  • Easy to use
  • Works across multiple temperatures and altitudes
  • 3M Catalog# TR-973
Specifications
Certifications/Compliance NIOSH Approved
Disposable Reusable
Product Type Airflow Indicator
For Use With (Equipment) TR-600 and TR-800 Series PAPR Systems
Model TR-973
Quantity 1 Ea.