missing translation for 'onlineSavingsMsg'
Learn More

Vermiculite, Solstice

Catalog No. 60046172
Change view
Click to view available options
Quantity:
10 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60046172 10 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60046172 Supplier Solstice Supplier No. 32977010KG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Tan chunks

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 1318-00-9
MDL Number MFCD00132837

Specifications

Melting Point 1000°C
Quantity 10 kg
Color Tan
Packaging Paper/Poly Bag
UN Number NONH for all modes of transport
Chemical Name or Material Vermiculite