missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VeriSpec™ Yttrium Standard for ICP 10 g/L in 5% HNO3, Ricca Chemical
Description
- 10 g/L in 5% HNO3
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Specifications
Specifications
| Packaging | Natural LDPE Bottle |
| CAS | 13494-98-9, 7697-37-2, 7732-18-5 |
| CAS Min % | 88.88%, 4.90%, 4.22% |
| CAS Max % | 92.50%, 5.10%, 4.40% |
| Specific Gravity | 1 |
| Physical Form | Liquid |
| Molecular Formula | H12N3O15Y |
| UN Number | UN3264 |
| Color | Colorless |
| pH | <2 |
| Show More |