missing translation for 'onlineSavingsMsg'
Learn More

Honeywell™ VeriShield™ 100 Series Hard Hat Mount Ear Muffs

Catalog No. 19085485
Change view
Click to view available options
Color:
Black
CSA Z94.2:
Class A
Dielectric:
Yes
Noise Reduction Rating (NRR):
25 dB
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color CSA Z94.2 Dielectric Noise Reduction Rating (NRR)
19085485 Black Class A Yes 25 dB
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19085485 Supplier Honeywell™ Supplier No. 1035201VS

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

VeriShield™ Hard Hat Ear Muffs mount to a variety of helmets and ensures protection against noise levels while allowing voices and signals to be heard more naturally.

  • Helmet mounted versions
  • Wide cup opening ideal for workers with large ears, or who wear hearing aids
  • Lightweight cushioned headband
  • Memory foam ear cushions
  • Oil-resistant cup surface withstands oily, heavy-work environments and frequent cleaning
  • Features Sound Management Technology (SMT) allows voices and signals to be heard more naturally
  • Designed for use with other PPE
  • Dielectric (models 19-085-484, 19-085-485, 19-085-486 only)

Specifications

Color Black
ANSI S3.1974 Yes
CSA Z94.2 Class A
Cushion Color Black
For Use With (Application) Construction, Electrical, Manufacturing, Oil and Gas, Mining, Steel and Metals, Welding, Agriculture
High Visibility No
Model VS120DH
Pairs per Package 1 Ea.
Noise Reduction Rating (NRR) 25 dB
Product Type VeriShield
Dielectric Yes
Material ABS
Style Hard Hat
Show More Show Less