missing translation for 'onlineSavingsMsg'
Learn More

Honeywell™ VeriShield™ 100 Series Over-the-Head Ear Muffs

Catalog No. 19085531
Change view
Click to view available options
CSA Z94.2:
Class A
Class AL
Color:
Black
High-visibility Yellow
Foldable:
No
Noise Reduction Rating (NRR):
26 dB
30 dB
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. CSA Z94.2 Color Foldable Noise Reduction Rating (NRR)
19085531 Class AL High-visibility Yellow No 30 dB
19085477 Class A Black No 26 dB
19085482 Class AL Black No 30 dB
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19085531 Supplier Honeywell™ Supplier No. 1035110VS

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

VeriShield™ Over-the-Head style ear muffs feature air flow control technology for maximum attenuation without increasing pressure on the head.

  • Comfortably fits a wide range of head sizes
  • Wide cup opening is ideal for workers with large ears, or who wear hearing aids
  • Lightweight cushioned headband
  • Memory foam ear cushions
  • Oil-resistant cup surface withstands heavy-work environments and frequent cleaning
  • Features Sound Management Technology (SMT) which improves employee safety by blocking harmful noise while allowing voices and signals to be heard more naturally
  • Designed for use with other PPE

Specifications

Color High-visibility Yellow
ANSI S3.1974 Yes
CSA Z94.2 Class AL
For Use With (Application) Construction, Manufacturing, Oil and Gas, Mining, Steel and Metals, Welding, Agriculture
High Visibility Yes
Model VS130HV
Pairs per Package 1 Ea.
Noise Reduction Rating (NRR) 30 dB
Product Type VeriShield
Dielectric No
Material ABS
Style Over-the-head
Foldable No
Show More Show Less