missing translation for 'onlineSavingsMsg'
Learn More

(-)-Verbenone 95.0+%, TCI America™

Catalog No. V007225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V007225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. V007225G Supplier TCI America Supplier No. V007225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1196-01-6
Molecular Formula C10H14O
Molecular Weight (g/mol) 150.22
MDL Number MFCD00001343
InChI Key DCSCXTJOXBUFGB-JGVFFNPUSA-N
Synonym levoverbenone, --verbenone, l-verbenone, 1s---verbenone, --2-pinen-4-one, verbenone, unii-2xp0j7754u, 1s,5s-4,6,6-trimethylbicyclo 3.1.1 hept-3-en-2-one, verbenone, l, bicyclo 3.1.1 hept-3-en-2-one, 4,6,6-trimethyl-, 1s,5s
PubChem CID 92874
ChEBI CHEBI:78316
IUPAC Name (1S,5S)-4,6,6-trimethylbicyclo[3.1.1]hept-3-en-2-one
SMILES CC1=CC(=O)[C@H]2C[C@@H]1C2(C)C

Specifications

Color Yellow
Quantity 25 g
Formula Weight 150.22
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material (-)-Verbenone
EINECSNumber (4)-0617
TSCA Yes