missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Venturi Tube Demonstration Apparatus

Catalog No. S15108
Change view
Click to view available options
Grade:
7 to 14
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S15108 7 to 14
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S15108 Supplier Eisco™ Supplier No. PH0137VD

Venturi Tube Demonstration Apparatus shows students the relationship between fluid velocity and pressure.

  • Venturi Tube demonstration apparatus, mounted on wooden base
  • Demonstrates the relationship between fluid velocity and pressure
  • Includes apparatus, 2 clamps, and instructional manual
  • Measures 12.75 in. tall and 17.75 in., wide
  • Great for classroom demonstrations

Required Materials (Not Included)

  • Water and/or fluids of various densities, food coloring, and paper towels
  • Air source and alcohol (optional)

Specifications

Product Type Venturi Tube Demonstration
Description Mounted on a Wooden Base
Grade 7 to 14
For Use With (Application) Investigating the Relationship Between Fluid Velocity and Pressure
Includes Apparatus, 2 Clamps, and Instruction Manual
Required Accessories Fluids in Various Densities, Food Coloring, Paper Towels
Dimensions (L x W x H) 45 x 13 x 32 cm