Learn More
Pyramex™ Venture II™ Safety Glasses
Catalog No. 19817370
Change view
Click to view available options
Frame Color:
Black
Lens Coating:
Anti-fog, Scratch-resistant, Impact-resistant, UV-treated
Scratch-resistant, Impact-resistant, UV-treated
Scratch-resistant, UV-treated, Impact-resistant
Lens Tint:
Amber
Clear
Gray
Indoor/Outdoor Mirror
Infinity Blue
Silver Mirror
Sun Block Bronze
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Color Lens Coating Lens Tint
19817370 Black Scratch-resistant, Impact-resistant, UV-treated Gray
19822691 Black Scratch-resistant, Impact-resistant, UV-treated Amber
19151450 Black Scratch-resistant, Impact-resistant, UV-treated Sun Block Bronze
19151448 Black Anti-fog, Scratch-resistant, Impact-resistant, UV-treated Gray
19817372 Black Scratch-resistant, UV-treated, Impact-resistant Silver Mirror
19151446 Black Anti-fog, Scratch-resistant, Impact-resistant, UV-treated Clear
19051451 Black Scratch-resistant, Impact-resistant, UV-treated Infinity Blue
19817374 Black Scratch-resistant, UV-treated, Impact-resistant Indoor/Outdoor Mirror
19817369 Black Scratch-resistant, UV-treated, Impact-resistant Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 19817370 Supplier Pyramex™ Supplier No. SB1820S
Venture II™ Safety Glasses feature full side protection without distracting sideshields or seams.
  • Lightweight spectacles integrate a sideshield into the lens
  • High-curve (9.75) dual-lens design provides more than 180° of coverage
  • Coated polycarbonate lenses provide superior scratch resistance and filter 99% of all harmful UV light
  • Temples adjust to four different sizes
  • Pillow-soft, thermoplastic elastomer ''nose-buds'' secure glasses on face for a comfortable fit
Compliance
Meet ANSI Z87.1 requirements and CSA Z94.3 standards
Specifications
Lens Coating Scratch-resistant, Impact-resistant, UV-treated
Certifications/Compliance Meet ANSI Z87.1 requirements and CSA Z94.3 standards
Lens Tint Gray
Product Type Safety Glasses
Frame Color Black
Temple Style Adjustable
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
Frame Material Polycarbonate, Nylon
UV Protection U6
Frame Type Half Frame
Autoclavable Autoclavable
Show More Show Less