missing translation for 'onlineSavingsMsg'
Learn More

Mopec Ventilated Tissue Storage System

Provides a safe and functional method for storing tissue samples. Mopec™ Ventilated Tissue Storage System is the practical answer to everyday tissue storage and transfer needs.

Supplier:  Mopec IS32401*

Catalog No. 22444190

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

  • All Stainless Steel cabinet construction
  • Door to the cabinet is clear heavy duty Lexan and may be hinged on the left or right
  • Key lock included
  • Topside ventilation duct makes it easy to adapt to most laboratory ventilation systems
  • Four leveling feet and an adjustable stainless steel floor section allows convenient and easy installation
  • During transport, smaller containers are conveniently located in trays which provide spill proof containment and a means of absorbing and neutralizing formalin
  • Ensures a sturdy stand alone cabinet; eliminating the wobbliness associated with a single cabinet
  • Equipped with a self adjusting floor section and a heavy duty, clear, door for convenient spill cleanup and easy viewing of contents
  • The portable, 5 tier, shelving rack conveniently stores within the cabinet for proper ventilation of its contents
  • This rack eliminates the need to transfer trays and samples to other mobile carts for intra-facility transport simply load specimens once; for both proper storage and future transport
  • The 5 diameter rubber casters allow for easy maneuverability throughout any hospital or laboratory facility
  • Easily adapts to most laboratory ventilation systems, requiring simple field hookups
  • A 6 diameter duct stub is located on top of the cabinet for convenience
  • Shipped fully assembled
  • Storage containers not included

Warranty

1 Year Warranty

Specifications

Specifications

Tissue Storage System
85 in.
30 in.
86.36 cm
Stainless Steel
34 in.
215.9 cm
76.2 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions