missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Vent Filter

Catalog No. TANKV01A1
Change view
Click to view available options
For Use With (Equipment):
High Flow Applications Storage Tank
Storage Tank
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
TANKV01A1 Storage Tank
TANKVH1A1 High Flow Applications Storage Tank
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. TANKV01A1 Supplier MilliporeSigma™ Supplier No. TANKV01A1
Only null left

Advanced protection against airborne contaminants for storage tanks. For use with Milli-Q™ IQ tanks.

The Vent Filter has been designed to provide an advanced protection against airborne contaminants for high Pure water stored in Milli-Q IQ storage tanks.

Specifications

For Use With (Equipment) Storage Tank
Type Vent Filter