Learn More
Brady™ Velocity™ 16 in. Compact Cable Lockout Padlocks, Keyed Different
Catalog No. 19029923
Change view
Click to view available options
Color:
Black
Blue
Brown
Green
Orange
Purple
Red
White
Yellow
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19029923 Yellow
19029891 Black
19029896 Blue
19029899 Brown
19029904 Green
19029907 Orange
19029910 Purple
19029915 Red
19029918 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 19029923 Supplier Brady™ Supplier No. CPPLYLW400CBKD
Only null left
Accelerate your program with Velocity™ Compact Cable Lockout Padlocks - engineered smaller and versatile for complex or unique lockout applications.
Available in the widest variety of colors, materials and styles, and ideal for any size lockout program.
Specifications
Color Yellow
Key Type Keyed Different
Material Nylon
Quantity 1 Ea.
Product Type Safety Padlock
Height (English) Shackle 8 in.