Recombinant Protein;1MG;Mouse VEGF R3, Mouse IgG2a Fc Tag, low endotoxin (FL4-M5251) is expressed from human 293 cells (HEK293). It contains AA Tyr 25 - Asp 770 (Accession # P35917-1).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 111.5 kDa. The protein migrates as 116-125 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human VEGF-C, His Tag (Cat. No. VEC-H4225) at 5 g/mL (100 L/well) can bind Mouse VEGF R3, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. FL4-M5251) with a linear range of 1-10 ng/mL (QC tested).