Learn More
Epredia™ Vega 8 Hopper Laser Cassette Printer with DUO and Total Care Package
Catalog No. 21403526
Change view
Click to view available options
Capacity:
640 Cassettes
Catalog No. Capacity
21403526 640 Cassettes
1 options
Catalog No. 21403526 Supplier Epredia™ Supplier No. 8000064B
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Produces high quality prints combined with a patented chemical additive on the cassette to deliver consistency and efficiency. Built in interface connectivity ensures LIS/LIM compatibility.
  • Laser printing technology for permanent cassette marking
  • Supports alphanumeric characters and barcode formats, including linear, bidimensional, and ASCII codes
  • High-resolution printing at 600 dpi for clear and precise output
  • Print speed of approximately 5 seconds per cassette
  • Designed for high-throughput workflows with automated cassette feeding
  • Suitable for high-volume laboratory environments
  • No consumables such as print heads or ribbons required
  • Reduced maintenance and lower operating costs
  • Available in two configurations: single cassette hopper (NOVA) and six-cassette hoppers (VEGA)
  • Supports multiple cassette colors and styles
  • Fully automated operation
  • Compact footprint for space-efficient installation
  • Low noise during operation
  • Includes 1-year base warranty with optional additional 2-year service plan
  • Service plan covers parts, labor, and annual preventive maintenance
  • Service agreement applicable when using compatible laser cassettes
  • Electrical requirements: 100/240 V, 50/60 Hz
  • Complies with IEC 61010-2-101:2015, IEC 61010-1:2010, IEC 61326-1:2012, IEC 61326-2-6:2018 standards
Specifications
Capacity 640 Cassettes
Includes DUO and Total Care Package
Type Laser Printer
Voltage 100/240 V
Color Gray