Learn More
Vecuronium bromide, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 461972500
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| Vecuronium bromide | |
| 98% | |
| 250 mg | |
| VEPSYABRBFXYIB-PWXDFCLTSA-M | |
| 1-[(1R,2S,3aS,3bR,5aS,7S,8S,9aS,9bS,11aS)-1,7-bis(acetyloxy)-9a,11a-dimethyl-8-(piperidin-1-yl)-hexadecahydro-1H-cyclopenta[a]phenanthren-2-yl]-1-methylpiperidin-1-ium bromide | |
| 637.75 |
| 50700-72-6 | |
| C34H57BrN2O4 | |
| Solubility aq. soln.: slightly soluble. Other solubilities: soluble in ethanol, dichloroethane, acetone | |
| [Br-].CC(=O)O[C@H]1[C@H](C[C@H]2[C@@H]3CC[C@H]4C[C@H](OC(C)=O)[C@H](C[C@]4(C)[C@H]3CC[C@]12C)N1CCCCC1)[N+]1(C)CCCCC1 | |
| 637.74 |
Chemical Identifiers
| 50700-72-6 | |
| 637.74 | |
| 1-[(1R,2S,3aS,3bR,5aS,7S,8S,9aS,9bS,11aS)-1,7-bis(acetyloxy)-9a,11a-dimethyl-8-(piperidin-1-yl)-hexadecahydro-1H-cyclopenta[a]phenanthren-2-yl]-1-methylpiperidin-1-ium bromide |
| C34H57BrN2O4 | |
| VEPSYABRBFXYIB-PWXDFCLTSA-M | |
| [Br-].CC(=O)O[C@H]1[C@H](C[C@H]2[C@@H]3CC[C@H]4C[C@H](OC(C)=O)[C@H](C[C@]4(C)[C@H]3CC[C@]12C)N1CCCCC1)[N+]1(C)CCCCC1 |
Safety and Handling
GHS H Statement Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
Wear protective gloves/protective clothing/eye protection/face protection.
GHS Signal Word: Warning
EINECSNumber : 256-723-9
RUO â Research Use Only