Learn More
Bullard™ Vector™ Helmet
Catalog No. 2705895
Change view
Click to view available options
Color:
Blue
Forest Green
Red
White
Suspension Type:
4-Point Ratchet
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Suspension Type
2705895 Forest Green 4-Point Ratchet
19088778C Blue 4-Point Ratchet
19088778A Red 4-Point Ratchet
19088778B White 4-Point Ratchet
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 2705895 Supplier Bullard™ Supplier No. VTFGR
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications
Certifications/Compliance ANSI Z89.1-1997 Type II G and E; CSA 294.1-1992 Class E
Color Forest Green
Material Shell: HDPE, Core: Polystyrene Foam
Suspension Type 4-Point Ratchet