Learn More
Vector Laboratories VECTASTAIN ANTI-MOUSE ABC-AP
Catalog No. NC9631600
Encompass
VECTASTAIN ABC-AP KIT (Mouse IgG),1 kit,consisting of 3mL of blocking serum,1mL of the biotinylated affinity-purified antibody or 2mL containing 2.1mg of the universal antibody,and 2mL each of the Avidin DH solution and biotinylated enzymes
Catalog No. NC9631600 Supplier Vector Laboratories Supplier No. AK5002
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
VECTASTAIN ABC-AP KIT (Mouse IgG),1 kit,consisting of 3mL of blocking serum,1mL of the biotinylated affinity-purified antibody or 2mL containing 2.1mg of the universal antibody,and 2mL each of the Avidin DH solution and biotinylated enzymes