Learn More
Eppendorf™ Varispenser™ 2x Bottle-Top Dispenser with Recirculation Valve
Catalog No. 05413559
Change view
Click to view available options
Volume (Metric):
0.2 to 2.0 mL
0.5 to 5.0 mL
1.0 to 10 mL
10 to 100 mL
2.5 to 25 mL
5.0 to 50 mL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
05413559 1.0 to 10 mL
05413557 0.2 to 2.0 mL
05413558 0.5 to 5.0 mL
05413560 2.5 to 25 mL
05413561 5.0 to 50 mL
05413562 10 to 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 05413559 Supplier Eppendorf™ Supplier No. 4967000030
Edge
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Ideal for dispensing aliquots of liquid from large supply bottles.
  • Recirculation valve
  • Standard GL 45 thread on all sizes for most common bottle threads
  • Safety valve balls in discharge valve prevents leakage when discharge tube is not mounted
  • Screwable air vent cap for easy assembly of drying tube or microfilter
  • Oval-shaped dispenser housing for easy transportation
  • PFA seal of the sliding piston prevents seizing up
  • Fixation slide on interior toothed track for rapid and accurate volume adjustment
  • High chemical resistance of all dispensing components
  • Telescopic aspirating tubes for variable adjustment to bottle height
  • Fully autoclavable without disassembly
  • User adjustment option
Specifications
Product Type Analog Bottletop Dispenser with Adapters
Channels 1
Volume (Metric) 1.0 to 10 mL
Includes Recirculation valve, adapters GL 25, GL 28/S 28, GL 32, GL 38, S 40, and telescopic intake tube (125 – 240 mm)