Learn More
Eppendorf™ Varispenser™ 2x Bottle-Top Dispenser with Recirculation Valve
Ideal for dispensing aliquots of liquid from large supply bottles.
Supplier:  Eppendorf™ 4967000022
 View more versions of this product
Catalog No. 05413558
Only null left
Description
Description
Ideal for dispensing aliquots of liquid from large supply bottles.
  • Recirculation valve
  • Standard GL 45 thread on all sizes for most common bottle threads
  • Safety valve balls in discharge valve prevents leakage when discharge tube is not mounted
  • Screwable air vent cap for easy assembly of drying tube or microfilter
  • Oval-shaped dispenser housing for easy transportation
  • PFA seal of the sliding piston prevents seizing up
  • Fixation slide on interior toothed track for rapid and accurate volume adjustment
  • High chemical resistance of all dispensing components
  • Telescopic aspirating tubes for variable adjustment to bottle height
  • Fully autoclavable without disassembly
  • User adjustment option
Specifications
Chemical Identifiers
1
Specifications
Analog Bottletop Dispenser with Adapters
PTFE, Polypropylene, Glass
Recirculation valve, adapters GL 25, GL 28/S 28, GL 32, GL 38, S 40, and telescopic intake tube (125 – 240 mm)
1
0.5 to 5.0 mL
Product Suggestions
Product Suggestions