missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Variamine Blue B 98.0+%, TCI America™
$63.38 - $206.90
Chemical Identifiers
| CAS | 3566-44-7 |
|---|---|
| Molecular Formula | C13H15ClN2O |
| Molecular Weight (g/mol) | 250.726 |
| MDL Number | MFCD00012989 |
| InChI Key | HPQQXLXIGHOKNZ-UHFFFAOYSA-N |
| PubChem CID | 77108 |
| IUPAC Name | 4-N-(4-methoxyphenyl)benzene-1,4-diamine;hydrochloride |
| SMILES | COC1=CC=C(C=C1)NC2=CC=C(C=C2)N.Cl |
Chemical Identifiers
| 3566-44-7 | |
| 250.726 | |
| HPQQXLXIGHOKNZ-UHFFFAOYSA-N | |
| 4-N-(4-methoxyphenyl)benzene-1,4-diamine;hydrochloride |
| C13H15ClN2O | |
| MFCD00012989 | |
| 77108 | |
| COC1=CC=C(C=C1)NC2=CC=C(C=C2)N.Cl |
Specifications
| 255°C | |
| C13H15ClN2O | |
| 4-Amino-4′C-methoxydiphenylamine Hydrochloride, N-(4-Methoxyphenyl)-1,4-phenylenediamine Hydrochloride, Variamine Blue B Hydrochloride | |
| COC1=CC=C(C=C1)NC2=CC=C(C=C2)N.Cl | |
| 250.726 | |
| 250.73 | |
| Blue | |
| Crystal/Powder |
| 3566-44-7 | |
| MFCD00012989 | |
| HPQQXLXIGHOKNZ-UHFFFAOYSA-N | |
| 4-N-(4-methoxyphenyl)benzene-1,4-diamine;hydrochloride | |
| 77108 | |
| ≥98.0% (T) | |
| 1 g | |
| Variamine Blue B [Redox Indicator] |
Safety and Handling
EINECSNumber : (5)-2320
TSCA : Yes