missing translation for 'onlineSavingsMsg'
Learn More

United Scientific™ Variable Volume, Fully Autoclavable Micropipettes

Catalog No. S43754
Change view
Click to view available options
Volume (Metric):
0.2 to 2.0 μL
0.5 to 10 μL
10 to 100 μL
100 to 1000 μL
2 to 10 mL
2 to 20 μL
20 to 200 μL
5 to 50 μL
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
S43754 2 to 10 mL
S43746 0.2 to 2.0 μL
S43747 0.5 to 10 μL
S43748 2 to 20 μL
S43749 5 to 50 μL
S43750 10 to 100 μL
S43751 20 to 200 μL
S43752 100 to 1000 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. S43754 Supplier United Scientific™ Supplier No. PRV10000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

These Variable Volume, Fully Autoclavable Micropipettes are highly accurate and easy to calibrate/maintain.

  • Built-in tip ejectors and autoclavable tip cones work on a click stop digital system.
  • Easy to disassemble prior to autoclaving.
  • Manufactured in an ISO 9001:2008 certified facility.
  • Pipettes are individually calibrated according to ISO 8655 standards.
  • A calibration certificate is provided with each pipette.

Specifications

Autoclavable Autoclavable
Calibrated Yes
Description Calibrated According to ISO 8655 Standards
Material Plastic
Sterility Non-sterile
For Use With (Application) For use with 10 mL tips
Graduated No
Graduations 100 μL
Graduation Type Digital
Includes Calibration Certificate
Color Blue
Volume (Metric) 2 to 10 mL
Length (Metric) 228 mm
Type Volumetric Pipet
Disposable Reusable
Show More Show Less