Learn More
Alkali Scientific VaporSeal™ Titan™ Microplate Sealing Starter Kit, Titan Dispenser + 2 Rolls of VaporSeal, 50 Sheets/Roll, Sterile
Catalog No. 502388956
Encompass
The dispenser facilitates the quick and easy dispensing of film from a roll allowing microplates to be sealed three times faster than using sheeted films. Environmentally friendly; no paper liner to remove and discard; utilizes approximately 59% less material than AeraSeal sheeted film. Each sheet is separated by an adhesive-free, perforated, color-coded red band. The smooth lower surface of the dispenser can be used to seal the film to the plate. Dispenser protects unused seals from contamination. Lower support prop prevents dispensing bill from contacting surfaces to prevent contamination. Replacement rolls can be easily installed by releasing the locking clasps on the dispenser.
Catalog No. 502388956 Supplier Alkali Scientific Supplier No. SF3VPS
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The dispenser facilitates the quick and easy dispensing of film from a roll allowing microplates to be sealed three times faster than using sheeted films. Environmentally friendly; no paper liner to remove and discard; utilizes approximately 59% less material than AeraSeal sheeted film. Each sheet is separated by an adhesive-free, perforated, color-coded red band. The smooth lower surface of the dispenser can be used to seal the film to the plate. Dispenser protects unused seals from contamination. Lower support prop prevents dispensing bill from contacting surfaces to prevent contamination. Replacement rolls can be easily installed by releasing the locking clasps on the dispenser.