Learn More
SOMNI Scientific™ Vaporizer Service Exchange Iso Tec 4 (Specify Fill Type, Mount)
Catalog No. 04777159
Change view
Click to view available options
Model:
Tec 4 Isoflurane
Catalog No. Model
04777159 Tec 4 Isoflurane
1 options
Catalog No. 04777159 Supplier SOMNI Scientific™ Supplier No. VX6022
Only null left
Maintains device accuracy, proper function and prevents exposure from leaks. A fully serviced and calibrated vaporizer of the same type is shipped to the customer. Customer uses prepaid return label to send their vaporizer back to our service center. No downtime.
  • Isoflurane, Tec 4
  • Specify Fill type, Mount and Serial number when ordering
  • Maintains accuracy
  • Prevents exposure from leaks
  • Full Service and Calibration
    • Certificate of Calibration included
  • Service interval based on manufacturer recommendation
  • Warranty provided
Specifications
For Use With (Application) Animal Inhalant Anesthesia
For Use With (Equipment) RM-17003, RM-17004, AMD-17004, TT-17000, OC-21003, OC-21004
Material Brass
Length (Metric) 14.5 cm
Width (Metric) 10.5 cm
Model Tec 4 Isoflurane
Includes Calibration Certificate
For Veterinary Use Only