Learn More
Chemglass Life Sciences Vapor Tube, Steam Duct, 45mm, 24/40 Joint, Compatible with Buchi Rotary Evaporators, Combi-Clip Not Included, 170mm Approx OAL
Supplier: Chemglass Inc CG131720
Vapor Tube, Steam Duct, 45mm, 24/40 Joint, Compatible with Buchi Rotary Evaporators, Combi-Clip Not Included, 170mm Approx OAL. Replacement Vapor tubes compatible with Buchi rotary evaporators. Thread is made for use with standard Combi-Clip. Available in the 4 listed sizes. NOT SUPPLIED WITH COMBI-CLIP.Dimension A:CG-1317-20: 45mm