Learn More
BUCHI Vapor Duct for Rotavapor
Vapor Duct for use with the Rotavapor.
Supplier:  BUCHI 11075728
 View more versions of this product
Catalog No. 01673001
Only null left
Description
Description
  • For glass assembly V and C
  • Includes Combi-Clip
Specifications
Chemical Identifiers
Vapor Duct with Combi-Clip
22 mm
Specifications
Vapor Duct with Combi-Clip
Rotavapor R-80
29/32
22 mm
Product Suggestions
Product Suggestions