Learn More
Synthware™ Vapor Duct
Catalog No. 31500627
Change view
Click to view available options
Product Type:
Ika style
Labtech style
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
31500627 Ika style
31500628 Labtech style
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 31500627 Supplier Synthware™ Supplier No. V222440
Precision ground and polished.
Specifications
Quantity 1 Ea.
Standard Taper Joint 24/40
Product Type Ika style
Material Borosilicate Glass
For Use With (Application) Organic Synthesis