missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Vancomycin hydrochloride, Molecular Biology Grade

Catalog No. p-7042509
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6279003 1 g
AAJ6279006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6279003 Supplier Thermo Scientific Chemicals Supplier No. J6279003
Only null left

Vancomycin hydrochloride (vancomycin HCl, CAS # 1404-93-9) is the hydrochloride salt of the bactericidal agent, vancomycin. | CAS: 1404-93-9 | C66H76Cl3N9O24 | 1485.72 g/mol

Inhibits bacterial mucopeptide synthesis.Vancomycin hydrochloride is used as an antiviral and anti bacterial. binds to bacterial cell wall. It exhibits potent activity against Gram positive bacteria and is highly effective against MRSA.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Vancomycin hydrochloride is a glycopeptide antibiotic that prevents cell wall synthesis
  • Originally obtained from the bacterial strain Streptomyces Orientalis
  • Vancomycin binds to D-alanyl-D-alanine motifs in cell wall precursors and leads to cell death
  • It is active against some penicillin-resistant bacteria

Application

  • Vancomycin hydrochloride is intended for use in the research of infections and for antibiotic resistance testing

Chemical Identifiers

CAS 1404-93-9
Molecular Formula C66H76Cl3N9O24
Molecular Weight (g/mol) 1485.72
MDL Number MFCD03613611,MFCD03613611
InChI Key LCTORFDMHNKUSG-UHFFFAOYNA-N
PubChem CID 124080918
IUPAC Name 48-({3-[(4-amino-5-hydroxy-4,6-dimethyloxan-2-yl)oxy]-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl}oxy)-22-(carbamoylmethyl)-5,15-dichloro-2,18,32,35,37-pentahydroxy-19-[4-methyl-2-(methylamino)pentanamido]-20,23,26,42,44-pentaoxo-7,13-dioxa-21,24,27,41,43-pentaazaoctacyclo[26.14.2.2³,⁶.2¹⁴,¹⁷.1⁸,¹².1²⁹,³³.0¹⁰,²⁵.0³⁴,³⁹]pentaconta-3,5,8(48),9,11,14,16,29(45),30,32,34,36,38,46,49-pentadecaene-40-carboxylic acid hydrochloride
SMILES Cl.CNC(CC(C)C)C(=O)NC1C(O)C2=CC(Cl)=C(OC3=CC4=CC(OC5=C(Cl)C=C(C=C5)C(O)C5NC(=O)C(NC(=O)C4NC(=O)C(CC(N)=O)NC1=O)C1=CC(=C(O)C=C1)C1=C(O)C=C(O)C=C1C(NC5=O)C(O)=O)=C3OC1OC(CO)C(O)C(O)C1OC1CC(C)(N)C(O)C(C)O1)C=C2

Specifications

Type Vancomycin Hydrochloride
Physical Form Powder
Color White
Quantity 1 g
Sensitivity Hygroscopic
Beilstein 3704657
Merck Index 149929
Solubility Information Soluble in water; Slightly soluble in methanol,ethanol and dimethylsulfoxide.
Formula Weight 1485.73
Grade Molecular Biology
Chemical Name or Material Vancomycin hydrochloride
Show More Show Less
Hazard Category Serious eye damage/eye irritation (category 2), Specific target organ toxicity after single exposure (category 3), Skin sensitisation (category 1), Skin corrosion/irritation (category 2)
Hazard Statement Causes serious eye irritation, May cause respiratory irritation, May cause an allergic skin reaction, Causes skin irritation
RTECSNumber YW4380000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only