missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlorhydrate de vancomycine, Fisher BioReagents
Spécifications
Spécifications
| Catégorie | Chlorhydrate de vancomycine |
| Nom ou substance chimique | Vancomycin Hydrochloride |
| Forme physique | Poudre/Solide |
| pH | 2,5 à 4,5 |
| Sources | Streptomyces orientalis |
| Emballage | Verre ambré |
| Puissance | > 900 U/mg |
| Eau | <10.0% |
| Couleur | Blanc |
| Quantité | 1 g |
| Afficher plus de résultats |
Sécurité et manipulation
| Hazard Statement | WARNING! |
| Precautionary Statement | Emergency Overview Irritating to eyes, respiratory system and skin. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Obtain medical attention. Do not induce vomiting. Do not induce vomiting. Obtain medical attention. NFPA Health:2 Flammability:0 Instability:1 |